missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-DL-methionine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6637206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-DL-methionine | |
C10H19NO4S | |
boc-dl-met-oh, boc-dl-methionine, methionine,n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl methionine, 2-tert-butoxy carbonyl amino-4-methylsulfanyl butanoic acid, 2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoic acid, n-alpha-t-boc-l-methionine, n-tert-butoxycarbonyl-d-methionine, tert-butoxycarbonyl-l-methionine, pubchem11999 | |
CSCCC(NC(=O)OC(C)(C)C)C(O)=O | |
249.33 | |
249.33 | |
5 g |
93000-03-4 | |
MFCD00190811 | |
IMUSLIHRIYOHEV-UHFFFAOYNA-N | |
2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid | |
102827 | |
98% | |
Solid |
Chemical Identifiers
93000-03-4 | |
249.33 | |
IMUSLIHRIYOHEV-UHFFFAOYNA-N | |
102827 | |
CSCCC(NC(=O)OC(C)(C)C)C(O)=O |
C10H19NO4S | |
MFCD00190811 | |
boc-dl-met-oh, boc-dl-methionine, methionine,n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl methionine, 2-tert-butoxy carbonyl amino-4-methylsulfanyl butanoic acid, 2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoic acid, n-alpha-t-boc-l-methionine, n-tert-butoxycarbonyl-d-methionine, tert-butoxycarbonyl-l-methionine, pubchem11999 | |
2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only