missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-DL-alanine, 97%

$71.08 - $211.27

Chemical Identifiers

CAS 3744-87-4
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00063124
InChI Key QVHJQCGUWFKTSE-UHFFFAOYSA-N
Synonym boc-dl-alanine, boc-dl-ala-oh, 2-tert-butoxycarbonyl amino propanoic acid, n-boc-dl-alanine, 2-n-boc-amino-propionic acid, n-tert-butoxycarbonyl alanine, 2-tert-butoxycarbonyl amino propanoic acid, 2-tert-butoxycarbonylamino-propionic acid, 2-tert-butoxycarbonylamino propanoic acid, 2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 268471
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6226614 View Documents Alfa Aesar™
H6226614
25g Each for $71.08
 
AAH6226622 View Documents Alfa Aesar™
H6226622
100g Each for $211.27
 
Specifications

Chemical Identifiers

3744-87-4
189.211
QVHJQCGUWFKTSE-UHFFFAOYSA-N
268471
CC(C(=O)O)NC(=O)OC(C)(C)C
C8H15NO4
MFCD00063124
boc-dl-alanine, boc-dl-ala-oh, 2-tert-butoxycarbonyl amino propanoic acid, n-boc-dl-alanine, 2-n-boc-amino-propionic acid, n-tert-butoxycarbonyl alanine, 2-tert-butoxycarbonyl amino propanoic acid, 2-tert-butoxycarbonylamino-propionic acid, 2-tert-butoxycarbonylamino propanoic acid, 2-tert-butoxy carbonyl amino propanoic acid
2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

25g
C8H15NO4
boc-dl-alanine, boc-dl-ala-oh, 2-tert-butoxycarbonyl amino propanoic acid, n-boc-dl-alanine, 2-n-boc-amino-propionic acid, n-tert-butoxycarbonyl alanine, 2-tert-butoxycarbonyl amino propanoic acid, 2-tert-butoxycarbonylamino-propionic acid, 2-tert-butoxycarbonylamino propanoic acid, 2-tert-butoxy carbonyl amino propanoic acid
CC(C(=O)O)NC(=O)OC(C)(C)C
189.211
189.21
N-Boc-DL-alanine
3744-87-4
MFCD00063124
QVHJQCGUWFKTSE-UHFFFAOYSA-N
2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
268471
97%