Please login to your online account to display your discounted pricing

N-BOC-Diethanolamine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-BOC-Diethanolamine
CAS 103898-11-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H19NO4
Formula Weight 205.26
Synonym tert-Butyl N, N-bis(2-hydroxyethyl)carbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC440750050

  SDS
thermo fisher scientific
AC440750050
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440750250

  SDS
thermo fisher scientific
AC440750250
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid or Oil
Assay ≥97.5%
Formula Weight 205.26
Quantity 5mL
Color Colorless
Density 1.085g/mL
CAS Max % 100.0
Chemical Name or Material N-BOC-Diethanolamine
Assay Percent Range 98%
CAS Min % 97.5
CAS 103898-11-9
Molecular Formula C9H19NO4
Synonym tert-Butyl N, N-bis(2-hydroxyethyl)carbamate
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.