Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-valinol, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-D-valinol
CAS 106391-87-1
Formula Weight 203.28
MDL Number MFCD00235960
Synonym N-(tert-Butoxycarbonyl)-D-valinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6114903

 
alfa aesar™
H6114903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6114906

 
alfa aesar™
H6114906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6114914

 
alfa aesar™
H6114914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.28
Chemical Name or Material N-Boc-D-valinol
Physical Form Liquid
Assay 95%
MDL Number MFCD00235960
CAS 106391-87-1
Synonym N-(tert-Butoxycarbonyl)-D-valinol
Refractive Index 1.4510
Density 0.995g/mL
Boiling Point 218°C
Color Pale yellow
Quantity 1g