missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-valinol, 95%

Manufacturer:  Alfa Aesar™ H6114914

 View more versions of this product

Catalog No. AAH6114914



Specifications

Specifications

N-Boc-D-valinol
C10H21NO3
boc-d-val-ol
1.4510
tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate
11241137
Liquid
0.995g/mL
Yellow
106391-87-1
MFCD00235960
OOQRRYDVICNJGC-QMMMGPOBSA-N
CC(C)C(CO)NC(=O)OC(C)(C)C
203.282
203.28
95%
218°C
25g

Chemical Identifiers

106391-87-1
203.282
OOQRRYDVICNJGC-QMMMGPOBSA-N
11241137
CC(C)C(CO)NC(=O)OC(C)(C)C
C10H21NO3
MFCD00235960
boc-d-val-ol
tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate
Documents