Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-tyrosine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-D-tyrosine
CAS 70642-86-3
Molecular Formula C14H19NO5
Formula Weight 281.3
MDL Number MFCD00063030
Beilstein 3085467
Synonym Boc-D-Tyr-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6144014

  SDS
alfa aesar™
H6144014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 281.3
Chemical Name or Material N-Boc-D-tyrosine
Molecular Formula C14H19NO5
Synonym Boc-D-Tyr-OH
Assay 95%
Quantity 25g
Beilstein 3085467
MDL Number MFCD00063030
CAS 70642-86-3