Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-tert-leucine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-D-tert-leucine
CAS 124655-17-0
Formula Weight 231.29
MDL Number MFCD00065575
Synonym Boc-D-Tle-OH; Boc-D-α-tert-butylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6247303

 
alfa aesar™
H6247303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6247306

 
alfa aesar™
H6247306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62473MD

 
alfa aesar™
H62473MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.29
MDL Number MFCD00065575
Physical Form Solid
Quantity 1g
Synonym Boc-D-Tle-OH; Boc-D-α-tert-butylglycine
CAS 124655-17-0
Chemical Name or Material N-Boc-D-tert-leucine
Assay 95%