Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-prolinol, 99%

$91.46 - $91.46

Chemical Identifiers

Chemical Name or Material N-Boc-D-prolinol
CAS 83435-58-9
Molecular Formula C10H19NO3
Formula Weight 201.27
MDL Number MFCD00040580
Synonym Boc-D-Pro-ol; N-(tert-Butoxycarbonyl)-D-prolinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1626803

  SDS
alfa aesar™
L1626803
1g Each for $91.46
Description & Specifications

Specifications

Formula Weight 201.27
Chemical Name or Material N-Boc-D-prolinol
CAS 83435-58-9
Assay 99%
MDL Number MFCD00040580
Synonym Boc-D-Pro-ol; N-(tert-Butoxycarbonyl)-D-prolinol
Melting Point 59° to 62°C
Quantity 1g
Optical Rotation +54° (c=5 in Methanol)
Recommended Storage 2° to 8°C
Molecular Formula C10H19NO3
Safety and Handling

2° to 8°C