missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-proline methyl ester, 95%

$37.01 - $434.88

Chemical Identifiers

CAS 73323-65-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00190828
InChI Key WVDGSSCWFMSRHN-MRVPVSSYSA-N
Synonym boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
PubChem CID 1277667
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62672MD View Documents Alfa Aesar™
H62672MD
250mg Each for $37.01
 
AAH6267203 View Documents Alfa Aesar™
H6267203
1g Each for $109.64
 
AAH6267206 View Documents Alfa Aesar™
H6267206
5g Each for $434.88
 
Description

Description

Applications
N-Boc-D-proline methyl ester is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Chemical Identifiers

73323-65-6
229.276
WVDGSSCWFMSRHN-MRVPVSSYSA-N
1277667
CC(C)(C)OC(=O)N1CCCC1C(=O)OC
C11H19NO4
MFCD00190828
boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate

Specifications

250mg
C11H19NO4
boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
CC(C)(C)OC(=O)N1CCCC1C(=O)OC
229.276
229.27
N-Boc-D-proline methyl ester
73323-65-6
MFCD00190828
WVDGSSCWFMSRHN-MRVPVSSYSA-N
1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
1277667
95%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C