missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-D-proline methyl ester, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6267206

 View more versions of this product

Catalog No. AAH6267206



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-D-proline methyl ester is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Specifications

N-Boc-D-proline methyl ester
C11H19NO4
5 g
Soluble in DMSO.
CC(C)(C)OC(=O)N1CCCC1C(=O)OC
229.276
229.27
73323-65-6
MFCD00190828
boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
WVDGSSCWFMSRHN-MRVPVSSYSA-N
1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
1277667
95%

Chemical Identifiers

73323-65-6
229.276
WVDGSSCWFMSRHN-MRVPVSSYSA-N
1277667
CC(C)(C)OC(=O)N1CCCC1C(=O)OC
C11H19NO4
MFCD00190828
boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions