missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-prolinal, 97%

$295.54 - $905.09

Chemical Identifiers

CAS 73365-02-3
Molecular Formula C10H17NO3
Formula Weight 199.25
MDL Number MFCD01321389
Synonym N-(tert-Butoxycarbonyl)-D-prolinal; (R)-(+)-1-Boc-pyrrolidine-2-carboxaldehyde
Chemical Name or Material N-Boc-D-prolinal
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2629403 SDS Alfa Aesar™
H2629403
1g Each for $295.54
AAH2629406 SDS Alfa Aesar™
H2629406
5g Each for $905.09
Specifications Description & Specifications

Chemical Identifiers

73365-02-3
199.25
N-(tert-Butoxycarbonyl)-D-prolinal; (R)-(+)-1-Boc-pyrrolidine-2-carboxaldehyde
C10H17NO3
MFCD01321389
N-Boc-D-prolinal

Specifications

73365-02-3
199.25
228°C
>110°C (230°F)
+88° (c=1 in Chloroform)
97%
C10H17NO3
MFCD01321389
1g
N-(tert-Butoxycarbonyl)-D-prolinal; (R)-(+)-1-Boc-pyrrolidine-2-carboxaldehyde
N-Boc-D-prolinal

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : −30° to −10°C