missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-phenylalanine methyl ester, 98%

Manufacturer:  Alfa Aesar™ H6346603

 View more versions of this product

Catalog No. AAH6346603


 

Description

Description

Applications
N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

N-Boc-D-phenylalanine methyl ester
98%
MFCD00672509
SDSWSVBXRBXPRL-GFCCVEGCSA-N
methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
637614
Solid
77119-84-7
C15H21NO4
Boc-D-Phe-OMe
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC
279.336
279.34
1g

Chemical Identifiers

77119-84-7
279.336
SDSWSVBXRBXPRL-GFCCVEGCSA-N
637614
C15H21NO4
MFCD00672509
Boc-D-Phe-OMe
SDS Documents