missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-phenylalanine methyl ester, 98%

$62.46 - $925.27

Chemical Identifiers

CAS 77119-84-7
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD00672509
InChI Key SDSWSVBXRBXPRL-GFCCVEGCSA-N
Synonym boc-d-phenylalanine methyl ester, boc-d-phe-ome, r-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-dphe-ome, n-tert-butoxycarbonyl-d-phenylalanine methyl ester, d-phenylalanine,n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate
PubChem CID 637614
IUPAC Name methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6346603 View Documents Alfa Aesar™
H6346603
1g Each for $62.46
 
AAH6346606 View Documents Alfa Aesar™
H6346606
5g Each for $231.32
 
AAH6346614 View Documents Alfa Aesar™
H6346614
25g Each for $925.27
 
Description

Description

Applications
N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

77119-84-7
279.336
SDSWSVBXRBXPRL-GFCCVEGCSA-N
637614
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC
C15H21NO4
MFCD00672509
boc-d-phenylalanine methyl ester, boc-d-phe-ome, r-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-dphe-ome, n-tert-butoxycarbonyl-d-phenylalanine methyl ester, d-phenylalanine,n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate
methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate

Specifications

77119-84-7
MFCD00672509
SDSWSVBXRBXPRL-GFCCVEGCSA-N
methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
637614
Solid
1g
C15H21NO4
boc-d-phenylalanine methyl ester, boc-d-phe-ome, r-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-dphe-ome, n-tert-butoxycarbonyl-d-phenylalanine methyl ester, d-phenylalanine,n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC
279.336
279.34
98%
N-Boc-D-phenylalanine methyl ester