Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-phenylalanine, 98%

$112.86 - $112.86

Chemical Identifiers

Chemical Name or Material N-Boc-D-phenylalanine
CAS 18942-49-9
Molecular Formula C14H19NO4
Formula Weight 265.31
MDL Number MFCD00063149
Beilstein 3593396
Synonym Boc-D-Phe-OH; N-(tert-Butoxycarbonyl)-D-phenylalanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0872206

  SDS
alfa aesar™
L0872206
5g Each for $112.86
Description & Specifications

Specifications

Formula Weight 265.31
CAS 18942-49-9
Chemical Name or Material N-Boc-D-phenylalanine
Assay 98%
Molecular Formula C14H19NO4
Synonym Boc-D-Phe-OH; N-(tert-Butoxycarbonyl)-D-phenylalanine
Optical Rotation −24.5° (c=1 in Ethanol)
MDL Number MFCD00063149
Recommended Storage 2° to 8°C
Beilstein 3593396
Quantity 5g
Melting Point 85° to 88°C
Safety and Handling

2° to 8°C