Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-phenylalanine 4-nitrophenyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-D-phenylalanine 4-nitrophenyl ester
CAS 16159-70-9
Formula Weight 386.4
MDL Number MFCD00069874
Synonym Boc-D-Phe-ONP; (R)-2-(Boc-amino)-3-phenylpropionic acid 4-nitrophenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6675806

  SDS
alfa aesar™
H6675806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6675814

  SDS
alfa aesar™
H6675814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 386.4
Chemical Name or Material N-Boc-D-phenylalanine 4-nitrophenyl ester
MDL Number MFCD00069874
CAS 16159-70-9
Synonym Boc-D-Phe-ONP; (R)-2-(Boc-amino)-3-phenylpropionic acid 4-nitrophenyl ester
Physical Form Solid
Assay 95%
Quantity 5g
Sensitivity Moisture Sensitive
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C