Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-norvaline dicyclohexylammonium salt, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-D-norvaline dicyclohexylammonium salt
CAS 57521-85-4
Formula Weight 398.58
MDL Number MFCD11041002
Synonym Boc-D-Nva-OH.DCHA; (R)-2-(Boc-amino)pentanoic acid dicyclohexylammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6179603

 
alfa aesar™
H6179603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6179606

 
alfa aesar™
H6179606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 398.58
Chemical Name or Material N-Boc-D-norvaline dicyclohexylammonium salt
Physical Form Powder
Assay 98%
MDL Number MFCD11041002
CAS 57521-85-4
Synonym Boc-D-Nva-OH.DCHA; (R)-2-(Boc-amino)pentanoic acid dicyclohexylammonium salt
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C