missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-norvaline dicyclohexylammonium salt, 98%

Manufacturer:  Alfa Aesar™ H6179606

 View more versions of this product

Catalog No. AAH6179606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Boc-D-norvaline dicyclohexylammonium salt
C10H19NO4
2r-2-tert-butoxycarbonyl amino pentanoic acid
CCCC(C(=O)O)NC(=O)OC(C)(C)C
217.265
398.58
98%
5g
57521-85-4
MFCD11041002
INWOAUUPYIXDHN-SSDOTTSWSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
6995000
Powder
White

Chemical Identifiers

57521-85-4
217.265
INWOAUUPYIXDHN-SSDOTTSWSA-N
6995000
CCCC(C(=O)O)NC(=O)OC(C)(C)C
C10H19NO4
MFCD11041002
2r-2-tert-butoxycarbonyl amino pentanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents