Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-methionine, 98+%

$38.95 - $125.91

Chemical Identifiers

Chemical Name or Material N-Boc-D-methionine
CAS 5241-66-7
Molecular Formula C10H19NO4S
Formula Weight 249.32
MDL Number MFCD00038256
Beilstein 4294122
Synonym Boc-D-Met-OH; N-(tert-Butoxycarbonyl)-D-methionine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0920703

 
alfa aesar™
L0920703
1g Each for $38.95

AAL0920706

 
alfa aesar™
L0920706
5g Each for $125.91
Description & Specifications

Specifications

Formula Weight 249.32
Chemical Name or Material N-Boc-D-methionine
Assay ≥98%
Melting Point 47° to 50°C
MDL Number MFCD00038256
CAS 5241-66-7
Beilstein 4294122
Quantity 1g
Optical Rotation +22° (c=1 in Methanol)
Molecular Formula C10H19NO4S
Synonym Boc-D-Met-OH; N-(tert-Butoxycarbonyl)-D-methionine
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C