Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-leucinol, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-D-leucinol
CAS 106930-51-2
Formula Weight 217.31
MDL Number MFCD00235931
Synonym (R)-N-(tert-Butoxycarbonyl)leucinol; (R)-2-Boc-amino-4-methyl-1-pentanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6249803

  SDS
alfa aesar™
H6249803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.31
Physical Form Liquid
Density 0.983g/mL
Synonym (R)-N-(tert-Butoxycarbonyl)leucinol; (R)-2-Boc-amino-4-methyl-1-pentanol
Refractive Index 1.4490
Color Colorless
Boiling Point 213°C
MDL Number MFCD00235931
Quantity 1g
Chemical Name or Material N-Boc-D-leucinol
CAS 106930-51-2
Assay 98%