Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-glutamic acid 5-cyclohexyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-D-glutamic acid 5-cyclohexyl ester
CAS 133464-27-4
Formula Weight 329.39
MDL Number MFCD00076929
Synonym Boc-D-Glu(ochex)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6647203

 
alfa aesar™
H6647203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6647206

 
alfa aesar™
H6647206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66472MD

 
alfa aesar™
H66472MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 329.39
Chemical Name or Material N-Boc-D-glutamic acid 5-cyclohexyl ester
Physical Form Solid
Assay 95%
MDL Number MFCD00076929
CAS 133464-27-4
Synonym Boc-D-Glu(ochex)-OH
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C