missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-glutamic acid 1-tert-butyl ester, 95%

$80.51 - $957.89

Chemical Identifiers

CAS 73872-71-6
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.355
MDL Number MFCD00076926
InChI Key YMOYURYWGUWMFM-SECBINFHSA-N
Synonym 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid
PubChem CID 7018746
IUPAC Name (4R)-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62320MD View Documents Alfa Aesar™
H62320MD
250mg Each for $80.51
 
AAH6232003 View Documents Alfa Aesar™
H6232003
1g Each for $239.47
 
AAH6232006 View Documents Alfa Aesar™
H6232006
5g Each for $957.89
 
Specifications

Chemical Identifiers

73872-71-6
303.355
YMOYURYWGUWMFM-SECBINFHSA-N
7018746
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OC(C)(C)C
C14H25NO6
MFCD00076926
4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid
(4R)-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid

Specifications

73872-71-6
MFCD00076926
YMOYURYWGUWMFM-SECBINFHSA-N
(4R)-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
7018746
Powder
Brown to White
N-Boc-D-glutamic acid 1-tert-butyl ester
C14H25NO6
4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OC(C)(C)C
303.355
303.35
95%
250mg
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold