missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-D-glutamic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H62320MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-D-glutamic acid 1-tert-butyl ester | |
Brown to White | |
MFCD00076926 | |
YMOYURYWGUWMFM-UHFFFAOYNA-N | |
5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid | |
7018746 | |
95% | |
Powder |
73872-71-6 | |
C14H25NO6 | |
boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester | |
CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C | |
303.36 | |
303.35 | |
250 mg |
Chemical Identifiers
73872-71-6 | |
303.36 | |
YMOYURYWGUWMFM-UHFFFAOYNA-N | |
7018746 |
C14H25NO6 | |
MFCD00076926 | |
boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester | |
CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only