Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-glutamic acid 1-benzyl ester, 99%

$397.78 - $397.78

Chemical Identifiers

Chemical Name or Material N-Boc-D-glutamic acid 1-benzyl ester
CAS 34404-30-3
Molecular Formula C17H23NO6
Formula Weight 337.37
MDL Number MFCD00038266
Synonym Boc-D-Glu-OBzl; N-tert-Butoxycarbonyl-D-glutamic acid 1-benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2194406

  SDS
alfa aesar™
B2194406
5g Each for $397.78
Description & Specifications

Specifications

Formula Weight 337.37
Synonym Boc-D-Glu-OBzl; N-tert-Butoxycarbonyl-D-glutamic acid 1-benzyl ester
Assay 99%
Molecular Formula C17H23NO6
Quantity 5g
MDL Number MFCD00038266
Chemical Name or Material N-Boc-D-glutamic acid 1-benzyl ester
CAS 34404-30-3