Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-cyclohexylglycinol, 98%

$326.97 - $326.97

Chemical Identifiers

Chemical Name or Material N-Boc-D-cyclohexylglycinol
CAS 188348-00-7
Molecular Formula C13H25NO3
Formula Weight 243.35
MDL Number MFCD04112590
Synonym (R)-2-(tert-Butoxycarbonylamino)-2-cyclohexylethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2741506

  SDS
alfa aesar™
H2741506
5g Each for $326.97
Description & Specifications

Specifications

Formula Weight 243.35
Chemical Name or Material N-Boc-D-cyclohexylglycinol
Assay 98%
Molecular Formula C13H25NO3
Melting Point 85° to 86°C
MDL Number MFCD04112590
CAS 188348-00-7
Quantity 5g
Synonym (R)-2-(tert-Butoxycarbonylamino)-2-cyclohexylethanol
Optical Rotation +12° (c=2 in CHCl3)