Learn More
N-Boc-D-beta-proline, 95%, Thermo Scientific Chemicals
$266.56 - $266.56
Chemical Identifiers
| CAS | 72925-16-7 |
|---|---|
| Molecular Formula | C10H17NO4 |
| Molecular Weight (g/mol) | 215.25 |
| MDL Number | MFCD03094727 |
| InChI Key | HRMRQBJUFWFQLX-UHFFFAOYNA-N |
| Synonym | r-1-n-boc-beta-proline, r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, r-1-boc-pyrrolidine-3-carboxylic acid, r-1-boc-3-carboxyl pyrrolidine, boc-l-beta-proline, n-boc-d-beta-proline, r-n-boc-pyrrolidine-3-carboxylic acid, r-1-n-boc-pyrrolidine-3-carboxylic acid, 3r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl ester, 3r |
| PubChem CID | 1512491 |
| SMILES | CC(C)(C)OC(=O)N1CCC(C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5782503
|
Thermo Scientific Chemicals
H5782503 |
1 g |
Each for $266.56
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in Dimethyl sulfoxide (DMSO).
Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
Chemical Identifiers
| 72925-16-7 | |
| 215.25 | |
| HRMRQBJUFWFQLX-UHFFFAOYNA-N | |
| 1512491 |
| C10H17NO4 | |
| MFCD03094727 | |
| r-1-n-boc-beta-proline, r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, r-1-boc-pyrrolidine-3-carboxylic acid, r-1-boc-3-carboxyl pyrrolidine, boc-l-beta-proline, n-boc-d-beta-proline, r-n-boc-pyrrolidine-3-carboxylic acid, r-1-n-boc-pyrrolidine-3-carboxylic acid, 3r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl ester, 3r | |
| CC(C)(C)OC(=O)N1CCC(C1)C(O)=O |
Specifications
| 72925-16-7 | |
| MFCD03094727 | |
| r-1-n-boc-beta-proline, r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, r-1-boc-pyrrolidine-3-carboxylic acid, r-1-boc-3-carboxyl pyrrolidine, boc-l-beta-proline, n-boc-d-beta-proline, r-n-boc-pyrrolidine-3-carboxylic acid, r-1-n-boc-pyrrolidine-3-carboxylic acid, 3r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl ester, 3r | |
| HRMRQBJUFWFQLX-UHFFFAOYNA-N | |
| CC(C)(C)OC(=O)N1CCC(C1)C(O)=O | |
| 215.25 | |
| 215.25 | |
| N-Boc-D-beta-proline |
| C10H17NO4 | |
| 1 g | |
| Soluble in Dimethyl sulfoxide (DMSO). | |
| −15° (c=0.8 in Chloroform) | |
| (3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid | |
| 1512491 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only