Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-aspartic acid, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-D-aspartic acid
CAS 62396-48-9
Formula Weight 233.22
MDL Number MFCD00798618
Synonym Boc-D-Asp-OH; N-(tert-Butoxycarbonyl)-D-aspartic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6282003

  SDS
alfa aesar™
H6282003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6282006

  SDS
alfa aesar™
H6282006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6282014

  SDS
alfa aesar™
H6282014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.22
Chemical Name or Material N-Boc-D-aspartic acid
Physical Form Solid
Assay 98%
MDL Number MFCD00798618
CAS 62396-48-9
Synonym Boc-D-Asp-OH; N-(tert-Butoxycarbonyl)-D-aspartic acid
Quantity 1g