missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-aspartic acid, 98%

Manufacturer:  Alfa Aesar™ H6282014

 View more versions of this product

Catalog No. AAH6282014


 

Specifications

Specifications

N-Boc-D-aspartic acid
98%
MFCD00798618
KAJBMCZQVSQJDE-RXMQYKEDSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
7021110
Solid
62396-48-9
C9H15NO6
Boc-D-Asp-OH; N-(tert-Butoxycarbonyl)-D-aspartic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O
233.22
233.22
25g

Chemical Identifiers

62396-48-9
233.22
KAJBMCZQVSQJDE-RXMQYKEDSA-N
7021110
C9H15NO6
MFCD00798618
Boc-D-Asp-OH; N-(tert-Butoxycarbonyl)-D-aspartic acid
SDS Documents