missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-D-aspartic acid 4-benzyl ester, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6200003

 View more versions of this product

Catalog No. AAH6200003



Description

Description

N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

N-Boc-D-aspartic acid 4-benzyl ester
C16H21NO6
1 g
boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
323.345
323.34
51186-58-4
MFCD00038255
2305471
SOHLZANWVLCPHK-GFCCVEGCSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
6994663
98%

Chemical Identifiers

51186-58-4
323.345
SOHLZANWVLCPHK-GFCCVEGCSA-N
6994663
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
C16H21NO6
MFCD00038255
boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only