Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-aspartic acid 1-tert-butyl ester, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-D-aspartic acid 1-tert-butyl ester
CAS 77004-75-2
Formula Weight 289.33
MDL Number MFCD00236827
Beilstein 3553120
Synonym Boc-D-Asp-OtBu
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6671603

  SDS
alfa aesar™
H6671603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6671606

  SDS
alfa aesar™
H6671606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66716MD

  SDS
alfa aesar™
H66716MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 289.33
CAS 77004-75-2
Chemical Name or Material N-Boc-D-aspartic acid 1-tert-butyl ester
MDL Number MFCD00236827
Assay 98%
Beilstein 3553120
Synonym Boc-D-Asp-OtBu
Storage Note 1 -30° to -10°C
Quantity 1g
Physical Form Solid
Safety and Handling

-30° to -10°C