Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-allo-isoleucine dicyclohexylammonium salt, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-D-allo-isoleucine dicyclohexylammonium salt
CAS 55780-92-2
Formula Weight 412.62
Synonym Boc-D-Allo-Ile-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6682403

 
alfa aesar™
H6682403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6682406

 
alfa aesar™
H6682406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66824MD

 
alfa aesar™
H66824MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 412.62
CAS 55780-92-2
Chemical Name or Material N-Boc-D-allo-isoleucine dicyclohexylammonium salt
Physical Form Solid
Assay 98%
Quantity 1g
Synonym Boc-D-Allo-Ile-OH.DCHA
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C