missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-D-alanine methyl ester, 98%

$31.01 - $118.28

Chemical Identifiers

CAS 91103-47-8
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00191865
InChI Key GJDICGOCZGRDFM-ZCFIWIBFSA-N
Synonym boc-d-ala-ome, boc-d-ala.ome, boc-d-alanine methyl ester, d-alanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino propanoate, n-boc-d-alanine methyl ester, n-t-boc-d-alanine methyl ester, n-tert-butoxycarbonyl-d-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino propanoate
PubChem CID 637612
IUPAC Name methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C(=O)OC)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6174203
View Documents
Alfa Aesar™
H6174203
1g Each for $31.01
 
AAH6174206
View Documents
Alfa Aesar™
H6174206
5g Each for $118.28
 
Specifications

Chemical Identifiers

91103-47-8
203.238
GJDICGOCZGRDFM-ZCFIWIBFSA-N
637612
CC(C(=O)OC)NC(=O)OC(C)(C)C
C9H17NO4
MFCD00191865
boc-d-ala-ome, boc-d-ala.ome, boc-d-alanine methyl ester, d-alanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino propanoate, n-boc-d-alanine methyl ester, n-t-boc-d-alanine methyl ester, n-tert-butoxycarbonyl-d-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino propanoate
methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate

Specifications

91103-47-8
MFCD00191865
boc-d-ala-ome, boc-d-ala.ome, boc-d-alanine methyl ester, d-alanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino propanoate, n-boc-d-alanine methyl ester, n-t-boc-d-alanine methyl ester, n-tert-butoxycarbonyl-d-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino propanoate
CC(C(=O)OC)NC(=O)OC(C)(C)C
203.238
203.24
98%
White
1g
N-Boc-D-alaninemethylester
C9H17NO4
4310313
GJDICGOCZGRDFM-ZCFIWIBFSA-N
methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
637612
Powder
1.030 g/mL
34°C to 37°C
113°C (235°F)
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures