missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-cis-4-hydroxy-D-proline methyl ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6313603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-cis-4-hydroxy-D-proline methyl ester | |
White | |
MFCD00797548 | |
MZMNEDXVUJLQAF-HTQZYQBOSA-N | |
1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate | |
6951202 | |
95% | |
Powder |
114676-69-6 | |
C11H19NO5 | |
2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome | |
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O | |
245.275 | |
245.27 | |
1 g |
Chemical Identifiers
114676-69-6 | |
245.275 | |
MZMNEDXVUJLQAF-HTQZYQBOSA-N | |
6951202 | |
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O |
C11H19NO5 | |
MFCD00797548 | |
2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome | |
1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only