Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-Arg-Val-Arg-Arg-7-amino-4-methylcoumarin

For cellular and molecular biology applications

Specifications

Product Type N-Boc-Arg-Val-Arg-Arg-7-amino-4-methylcoumarin
Molecular Weight 843.00
Storage Requirements -30° to -10°C
Sequence Boc-R-V-R-R-AMC
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66336LB0

 
alfa aesar™
J66336LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66336MCR

 
alfa aesar™
J66336MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type N-Boc-Arg-Val-Arg-Arg-7-amino-4-methylcoumarin
Molecular Weight 843.00
Storage Requirements -30° to -10°C
Sequence Boc-R-V-R-R-AMC
Format Solid
Regulatory Status RUO
Purification Method HPLC