missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-aniline, 97%

Manufacturer:  Alfa Aesar™ H5523206

 View more versions of this product

Catalog No. AAH5523206



Specifications

Specifications

N-Boc-aniline
133°C to 137°C
3422-01-3
C6H5NHCO2C(CH3)3
carbamic acid, phenyl-, 1,1-dimethylethyl ester
CC(C)(C)OC(=O)NC1=CC=CC=C1
193.25
193.25
97%
Brown to White
5g
C11H15NO2
MFCD00043723
KZZHPWMVEVZEFG-UHFFFAOYSA-N
tert-butyl N-phenylcarbamate
137930
Solid

Chemical Identifiers

3422-01-3
193.25
KZZHPWMVEVZEFG-UHFFFAOYSA-N
137930
CC(C)(C)OC(=O)NC1=CC=CC=C1
C11H15NO2
MFCD00043723
carbamic acid, phenyl-, 1,1-dimethylethyl ester
tert-butyl N-phenylcarbamate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents