Please login to your online account to display your discounted pricing

N-BOC-Amino-(4-hydroxycyclohexyl)carboxylic acid, 95%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC364615000

 View more versions of this product

Catalog No. AC364615000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥94%
Formula Weight 259.3
Quantity 500mg
Color White to off-white
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material N-BOC-Amino-(4-hydroxycyclohexyl)carboxylic acid
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C12H21NO5
MDL Number MFCD02683153
CAS 369403-08-7
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-BOC-Amino-(4-hydroxycyclohexyl)carboxylic acid
CAS 369403-08-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H21NO5
Formula Weight 259.3
MDL Number MFCD02683153
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents