Learn More
N-BOC-Allylamine, 98%, Thermo Scientific Chemicals
$111.48 - $111.48
Chemical Identifiers
CAS | 78888-18-3 |
---|---|
Molecular Formula | C8H15NO2 |
Molecular Weight (g/mol) | 157.21 |
InChI Key | AWARHXCROCWEAK-UHFFFAOYSA-N |
Synonym | tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine |
PubChem CID | 4067274 |
IUPAC Name | tert-butyl N-prop-2-enylcarbamate |
SMILES | CC(C)(C)OC(=O)NCC=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC434770100
|
Thermo Scientific Chemicals
434770100 |
10 g | Glass Bottle |
Each for $111.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78888-18-3 | |
157.21 | |
tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine | |
tert-butyl N-prop-2-enylcarbamate |
C8H15NO2 | |
AWARHXCROCWEAK-UHFFFAOYSA-N | |
4067274 | |
CC(C)(C)OC(=O)NCC=C |
Specifications
34.9°C to 38.3°C | |
48.0°C to 50.0°C (1.0mmHg) | |
100.0 | |
97.5% min. (GC) | |
Authentic | |
tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine | |
CC(C)(C)OC(=O)NCC=C | |
157.21 | |
157.21 | |
10 g | |
Low Melting Solid |
78888-18-3 | |
97.5 | |
Colorless | |
C8H15NO2 | |
Glass Bottle | |
AWARHXCROCWEAK-UHFFFAOYSA-N | |
tert-butyl N-prop-2-enylcarbamate | |
4067274 | |
98% | |
79°C | |
N-BOC-Allylamine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only