Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-5-methoxy-7-methylindole, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-5-methoxy-7-methylindole
CAS 1387445-50-2
Formula Weight 261.32
Synonym tert-Butyl 5-methoxy-7-methylindole-1-carboxylate; 5-Methoxy-7-methylindole-1-carboxylic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6390503

 
alfa aesar™
H6390503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6390506

 
alfa aesar™
H6390506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63905MD

 
alfa aesar™
H63905MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 261.32
Quantity 1g
Assay 97%
CAS 1387445-50-2
Physical Form Solid
Chemical Name or Material N-Boc-5-methoxy-7-methylindole
Synonym tert-Butyl 5-methoxy-7-methylindole-1-carboxylate; 5-Methoxy-7-methylindole-1-carboxylic acid tert-butyl ester