missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-BOC-4-Piperidone, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 330270050

 View more versions of this product

Catalog No. AC330270050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
199.25
White to slightly yellow
100.0
Glass bottle
99%
79099-07-3
Authentic
Crystalline Powder
5g
73° to 77°C
N-(tert-Butoxycarbonyl)-4-piperidone
C10H17NO3
N-BOC-4-Piperidone
98.5

Chemical Identifiers

79099-07-3
100.0
C10H17NO3
N-(tert-Butoxycarbonyl)-4-piperidone
≥98.5%
98.5
99%
199.25
N-BOC-4-Piperidone

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes skin irritation. Harmful if swallowed. Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning