Learn More
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 330271000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-BOC-4-Piperidone | |
98.5 | |
74.0°C to 75.0°C | |
Authentic | |
Glass bottle | |
MFCD00151800 | |
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone | |
ROUYFJUVMYHXFJ-UHFFFAOYSA-N | |
tert-butyl 4-oxopiperidine-1-carboxylate | |
735900 | |
99% |
79099-07-3 | |
100.0 | |
White to Yellow | |
98.5% min. (GC) | |
C10H17NO3 | |
100 g | |
Solubility in water: slightly soluble | |
CC(C)(C)OC(=O)N1CCC(=O)CC1 | |
199.25 | |
199.25 | |
Crystalline Powder |
Chemical Identifiers
79099-07-3 | |
199.25 | |
ROUYFJUVMYHXFJ-UHFFFAOYSA-N | |
735900 | |
CC(C)(C)OC(=O)N1CCC(=O)CC1 |
C10H17NO3 | |
MFCD00151800 | |
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone | |
tert-butyl 4-oxopiperidine-1-carboxylate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO – Research Use Only