missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-phenyl-L-phenylalanine, 95%

$29.45 - $92.68

Chemical Identifiers

CAS 147923-08-8
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.407
MDL Number MFCD00191184
InChI Key NBVVKAUSAGHTSU-KRWDZBQOSA-N
Synonym boc-l-4,4'-biphenylalanine, boc-4-phenyl-phe-oh, boc-3-4-biphenylyl-l-alanine, boc-bip-oh, boc-4-phenyl-l-phenylalanine, s-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-boc-l-biphenylalanine, boc-4-biphenyl-l-ala, boc-l-4,4'-biphenylphenylalanine, n-boc-4-phenyl-l-phenylalanine
PubChem CID 2761796
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C2=CC=CC=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62896MD View Documents Alfa Aesar™
H62896MD
250mg Each for $29.45
 
AAH6289603 View Documents Alfa Aesar™
H6289603
1g Each for $92.68
 
Specifications

Chemical Identifiers

147923-08-8
341.407
NBVVKAUSAGHTSU-KRWDZBQOSA-N
2761796
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C2=CC=CC=C2)C(=O)O
C20H23NO4
MFCD00191184
boc-l-4,4'-biphenylalanine, boc-4-phenyl-phe-oh, boc-3-4-biphenylyl-l-alanine, boc-bip-oh, boc-4-phenyl-l-phenylalanine, s-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-boc-l-biphenylalanine, boc-4-biphenyl-l-ala, boc-l-4,4'-biphenylphenylalanine, n-boc-4-phenyl-l-phenylalanine
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylphenyl)propanoic acid

Specifications

+21° (c=2 in Methanol)
250mg
C20H23NO4
boc-l-4,4'-biphenylalanine, boc-4-phenyl-phe-oh, boc-3-4-biphenylyl-l-alanine, boc-bip-oh, boc-4-phenyl-l-phenylalanine, s-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-boc-l-biphenylalanine, boc-4-biphenyl-l-ala, boc-l-4,4'-biphenylphenylalanine, n-boc-4-phenyl-l-phenylalanine
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C2=CC=CC=C2)C(=O)O
341.407
341.4
N-Boc-4-phenyl-L-phenylalanine
103°C to 107°C
147923-08-8
MFCD00191184
NBVVKAUSAGHTSU-KRWDZBQOSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylphenyl)propanoic acid
2761796
95%
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C