Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-oxo-L-proline, 98%

$84.69 - $703.66

Chemical Identifiers

Chemical Name or Material N-Boc-4-oxo-L-proline
CAS 84348-37-8
Molecular Formula C10H15NO5
Formula Weight 229.24
MDL Number MFCD01860669
Synonym (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2710403

  SDS
alfa aesar™
H2710403
1g Each for $212.46

AAH2710406

  SDS
alfa aesar™
H2710406
5g Each for $703.66

AAH27104MD

  SDS
alfa aesar™
H27104MD
250mg Each for $84.69
Description & Specifications

Specifications

Formula Weight 229.24
Chemical Name or Material N-Boc-4-oxo-L-proline
Assay 98%
Molecular Formula C10H15NO5
Melting Point ≈160°C
MDL Number MFCD01860669
CAS 84348-37-8
Quantity 1g
Synonym (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C

Applications
N-Boc-4-oxo-L-proline is used in practical Synthesis of Boc-Protected cis-4-Trifluoromethyl and cis-4-Difluoromethyl-l- prolines.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

2° to 8°C