missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-4-nitro-D-phenylalanine, 98%, Thermo Scientific™
$112.49 - $410.77
Chemical Identifiers
CAS | 61280-75-9 |
---|---|
Molecular Formula | C14H18N2O6 |
Molecular Weight (g/mol) | 310.31 |
MDL Number | MFCD00069880 |
InChI Key | XBQADBXCNQPHHY-UHFFFAOYNA-N |
Synonym | boc-4-nitro-d-phenylalanine, boc-d-phe 4-no2-oh, boc-d-4-nitrophe, boc-d-4-no2-phe-oh, r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, n-tert-butoxycarbonyl-4-nitro-d-phenylalanine, boc-d-4-nitrophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-4-nitro, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-4-nitrophenyl propanoic acid |
PubChem CID | 2755957 |
IUPAC Name | 2-{[(tert-butoxy)carbonyl]amino}-3-(4-nitrophenyl)propanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6166903
|
Thermo Scientific Chemicals
H6166903 |
1 g |
Each for $112.49
|
|
|||||
AAH6166906
|
Thermo Scientific Chemicals
H6166906 |
5 g |
Each for $410.77
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
61280-75-9 | |
310.31 | |
XBQADBXCNQPHHY-UHFFFAOYNA-N | |
2755957 | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O |
C14H18N2O6 | |
MFCD00069880 | |
boc-4-nitro-d-phenylalanine, boc-d-phe 4-no2-oh, boc-d-4-nitrophe, boc-d-4-no2-phe-oh, r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, n-tert-butoxycarbonyl-4-nitro-d-phenylalanine, boc-d-4-nitrophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-4-nitro, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-4-nitrophenyl propanoic acid | |
2-{[(tert-butoxy)carbonyl]amino}-3-(4-nitrophenyl)propanoic acid |
Specifications
61280-75-9 | |
C14H18N2O6 | |
1 g | |
boc-4-nitro-d-phenylalanine, boc-d-phe 4-no2-oh, boc-d-4-nitrophe, boc-d-4-no2-phe-oh, r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, n-tert-butoxycarbonyl-4-nitro-d-phenylalanine, boc-d-4-nitrophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-4-nitro, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-4-nitrophenyl propanoic acid | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O | |
310.31 | |
310.3 | |
N-Boc-4-nitro-D-phenylalanine |
115°C | |
MFCD00069880 | |
4203814 | |
XBQADBXCNQPHHY-UHFFFAOYNA-N | |
2-{[(tert-butoxy)carbonyl]amino}-3-(4-nitrophenyl)propanoic acid | |
2755957 | |
98% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold