Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-methylene-L-proline, 97%

$166.24 - $1,402.28

Chemical Identifiers

Chemical Name or Material N-Boc-4-methylene-L-proline
CAS 84348-38-9
Molecular Formula C11H17NO4
Formula Weight 227.26
MDL Number MFCD01861787
Synonym (S)-4-Methylenepyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2774603

  SDS
alfa aesar™
H2774603
1g Each for $417.15

AAH2774606

  SDS
alfa aesar™
H2774606
5g Each for $1,402.28

AAH27746MD

  SDS
alfa aesar™
H27746MD
250mg Each for $166.24
Description & Specifications

Specifications

Formula Weight 227.26
Chemical Name or Material N-Boc-4-methylene-L-proline
Assay 97%
Molecular Formula C11H17NO4
Melting Point 110° to 114°C
MDL Number MFCD01861787
CAS 84348-38-9
Quantity 1g
Synonym (S)-4-Methylenepyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
Recommended Storage 2° to 8°C

Applications
As important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry. The fluorous prolins were synthesized from commercially available N-Boc-4-methylene- L-proline.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

2° to 8°C