missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-BOC-4-methylene-L-proline, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 84348-38-9 |
---|---|
Molecular Formula | C11H17NO4 |
Molecular Weight (g/mol) | 227.26 |
MDL Number | MFCD01861787 |
InChI Key | ULLGRIBXGPATMA-UHFFFAOYNA-N |
Synonym | n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline |
PubChem CID | 21727417 |
IUPAC Name | (2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
SMILES | CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC449260010
|
Thermo Scientific Chemicals
449260010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC449260050
|
Thermo Scientific Chemicals
449260050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
84348-38-9 | |
227.26 | |
ULLGRIBXGPATMA-UHFFFAOYNA-N | |
21727417 | |
CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O |
C11H17NO4 | |
MFCD01861787 | |
n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline | |
(2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Specifications
84348-38-9 | |
96% min. (HPLC) | |
C11H17NO4 | |
1g | |
Solubility in water: insoluble. | |
CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O | |
227.26 | |
227.26 | |
N-BOC-4-methylene-L-proline |
Authentic | |
Glass Bottle | |
MFCD01861787 | |
n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline | |
ULLGRIBXGPATMA-UHFFFAOYNA-N | |
(2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
21727417 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only