Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-iodoaniline, 95%

$29.53 - $60.72

Chemical Identifiers

Chemical Name or Material N-Boc-4-iodoaniline
CAS 159217-89-7
Molecular Formula C11H14INO2
Formula Weight 319.14
MDL Number MFCD03490497
Synonym tert-Butyl 4-iodophenylcarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2783903

  SDS
alfa aesar™
H2783903
1g Each for $60.72

AAH27839MD

  SDS
alfa aesar™
H27839MD
250mg Each for $29.53
Description & Specifications

Specifications

Formula Weight 319.14
Melting Point 142° to 144°C
Chemical Name or Material N-Boc-4-iodoaniline
Synonym tert-Butyl 4-iodophenylcarbamate
CAS 159217-89-7
Molecular Formula C11H14INO2
MDL Number MFCD03490497
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Assay 95%
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.