Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-iodo-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-4-iodo-D-phenylalanine
CAS 176199-35-2
Formula Weight 391.21
MDL Number MFCD00079670
Beilstein 7518620
Synonym Boc-D-Phe(4-I)-OH; (R)-2-(Boc-amino)-3-(4-iodophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6662206

 
alfa aesar™
H6662206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6662214

 
alfa aesar™
H6662214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 391.21
Chemical Name or Material N-Boc-4-iodo-D-phenylalanine
MDL Number MFCD00079670
CAS 176199-35-2
Physical Form Powder
Synonym Boc-D-Phe(4-I)-OH; (R)-2-(Boc-amino)-3-(4-iodophenyl)propionic acid
Quantity 5g
Sensitivity Light Sensitive
Beilstein 7518620
Color White
Assay 95%