missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-BOC-4-Hydroxypiperidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC368271000

 View more versions of this product

Catalog No. AC368271000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-BOC-4-Hydroxypiperidine
Glass bottle
60.0°C to 65.0°C
97%
96.0
96% min. (GC)
MFCD01075174
PWQLFIKTGRINFF-UHFFFAOYSA-N
tert-butyl 4-hydroxypiperidine-1-carboxylate
643502
Powder
Authentic
White to Cream
100g
109384-19-2
100.0
C10H19NO3
1-boc-4-hydroxy-piperidine
CC(C)(C)OC(=O)N1CCC(CC1)O
201.27
201.27
97%

Chemical Identifiers

109384-19-2
201.27
PWQLFIKTGRINFF-UHFFFAOYSA-N
643502
CC(C)(C)OC(=O)N1CCC(CC1)O
C10H19NO3
MFCD01075174
1-boc-4-hydroxy-piperidine
tert-butyl 4-hydroxypiperidine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents