missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-4-(Fmoc-amino)-L-phenylalanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6354303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-4-(Fmoc-amino)-L-phenylalanine | |
C29H30N2O6 | |
boc-4-fmoc-amino-l-phenylalanine, boc-p-amino-phe fmoc-oh, boc-l-4-fmoc aminophenylalanine, boc-phe 4-nhfmoc-oh, boc-l-4-aminophenylalanine fmoc, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine, boc-d-4-fmoc aminophenylalanine, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O | |
502.567 | |
502.56 | |
1 g |
114346-31-5 | |
MFCD00151889 | |
ZKSJJSOHPQQZHC-VWLOTQADSA-N | |
(2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2755959 | |
95% | |
Solid |
Chemical Identifiers
114346-31-5 | |
502.567 | |
ZKSJJSOHPQQZHC-VWLOTQADSA-N | |
2755959 | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O |
C29H30N2O6 | |
MFCD00151889 | |
boc-4-fmoc-amino-l-phenylalanine, boc-p-amino-phe fmoc-oh, boc-l-4-fmoc aminophenylalanine, boc-phe 4-nhfmoc-oh, boc-l-4-aminophenylalanine fmoc, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine, boc-d-4-fmoc aminophenylalanine, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine | |
(2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only