missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-(Fmoc-amino)-L-phenylalanine, 95%

Manufacturer:  Alfa Aesar™ H6354306

 View more versions of this product

Catalog No. AAH6354306



Specifications

Specifications

114346-31-5
MFCD00151889
ZKSJJSOHPQQZHC-VWLOTQADSA-N
(2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
2755959
Solid
5g
C29H30N2O6
boc-4-fmoc-amino-l-phenylalanine, boc-d-4-fmoc aminophenylalanine, boc-l-4-aminophenylalanine fmoc, boc-l-4-fmoc aminophenylalanine, boc-p-amino-phe fmoc-oh, boc-phe 4-nhfmoc-oh, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
502.567
502.56
95%

Chemical Identifiers

114346-31-5
502.567
ZKSJJSOHPQQZHC-VWLOTQADSA-N
2755959
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
C29H30N2O6
MFCD00151889
boc-4-fmoc-amino-l-phenylalanine, boc-d-4-fmoc aminophenylalanine, boc-l-4-aminophenylalanine fmoc, boc-l-4-fmoc aminophenylalanine, boc-p-amino-phe fmoc-oh, boc-phe 4-nhfmoc-oh, n-1,1-dimethylethoxy carbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino-l-phenylalanine, n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-phenylalanine, s-3-4-9h-fluoren-9-yl methoxy carbonyl amino phenyl-2-tert-butoxycarbonyl amino propanoic acid, s-boc-4-fmoc-amino-phenylalanine
(2S)-3-[4-(9H-fluoren-9-ylmethoxycarbonylamino)phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents