missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ N-Boc-4-fluoro-L-phenylalanine, 95%
Manufacturer: Alfa Aesar™ H6137003
Specifications
N-Boc-4-fluoro-L-phenylalanine | |
C14H18FNO4 | |
4323475 | |
RCXSXRAUMLKRRL-NSHDSACASA-N | |
(2S)-3-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
7020910 | |
Powder | |
White | |
1g |
41153-30-4 | |
MFCD00079672 | |
boc-4-fluoro-l-phe-oh | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)F)C(=O)O | |
283.299 | |
283.3 | |
95% | |
83°C to 86°C |
Chemical Identifiers
41153-30-4 | |
283.299 | |
RCXSXRAUMLKRRL-NSHDSACASA-N | |
7020910 | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)F)C(=O)O |
C14H18FNO4 | |
MFCD00079672 | |
boc-4-fluoro-l-phe-oh | |
(2S)-3-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |